CH6371379239 Chem Impex International, Inc.

Na-Fmoc-L-histidine

  • MDL number MFCD00190885
  • Cas Number : 116611-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-His-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-l-histidine/3764?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-l-histidine/3764?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart