CH6371379240 Chem Impex International, Inc.

Fmoc-L-homocitrulline

  • MDL number MFCD00237387
  • Cas Number : 201485-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-HomoCit-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to pale yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-homocitrulline/3765?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-homocitrulline/3765?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart