CH6371379243 Chem Impex International, Inc.

Fmoc-Ile-Pro-OH

  • MDL number MFCD00237659
  • Cas Number : 109425-47-0
  • Molecular Weight : 450.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-ile-pro-oh/3768?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-ile-pro-oh/3768?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart