CH6371379245 Chem Impex International, Inc.

Fmoc-L-threoninol

  • MDL number MFCD00270218
  • Cas Number : 176380-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 91-98� C
Synonym Fmoc-L-Thr-ol
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-threoninol/3770?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-threoninol/3770?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart