CH6371379249 Chem Impex International, Inc.

Na-Fmoc-Ne-2,4-dinitrophenyl-L-lysine

  • MDL number MFCD00273454
  • Cas Number : 148083-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-160
Synonym Fmoc-L-Lys(Dnp)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-2-4-dinitrophenyl-l-lysine/3774?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-2-4-dinitrophenyl-l-lysine/3774?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart