CH6371379250 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-formyl-L-lysine

  • MDL number MFCD00237025
  • Cas Number : 201004-23-1
  • Molecular Weight : 396.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-143 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-formyl-l-lysine/3775?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-formyl-l-lysine/3775?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart