CH6371379252 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-palmitoyl-L-lysine

  • MDL number MFCD00237026
  • Cas Number : 201004-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-palmitoyl-l-lysine/3777?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-palmitoyl-l-lysine/3777?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart