CH6371379261 Chem Impex International, Inc.

Fmoc-4-amino-L-phenylalanine

  • MDL number MFCD00237664
  • Cas Number : 95753-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-205
Synonym Fmoc-L-Phe(4-NH2)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white to light yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-amino-l-phenylalanine/3788?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-amino-l-phenylalanine/3788?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart