CH6371379263 Chem Impex International, Inc.

Fmoc-4-fluoro-DL-phenylalanine

  • MDL number MFCD00237667
  • Cas Number : 264276-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-188
Synonym Fmoc-DL-Phe(4-F)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-fluoro-dl-phenylalanine/3790?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-fluoro-dl-phenylalanine/3790?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart