CH6371379264 Chem Impex International, Inc.

Fmoc-L-proline 4-nitrophenyl ester

  • MDL number MFCD00038532
  • Cas Number : 71989-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-120° C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Light yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-proline-4-nitrophenyl-ester/3791?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-proline-4-nitrophenyl-ester/3791?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart