CH6371379273 Chem Impex International, Inc.

DL-alpha-Methylleucine

  • MDL number MFCD00038403
  • Cas Number : 144-24-1
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-alpha-methylleucine/3800?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-alpha-methylleucine/3800?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart