CH6371379280 Chem Impex International, Inc.

DL-a-Methylproline hydrobromide

  • MDL number MFCD01632056
  • Cas Number : 1346136-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym alpha-Me-DL-Pro-OH HBr
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow to brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dl-alpha-methylproline-hydrobromide/3807?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dl-alpha-methylproline-hydrobromide/3807?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart