CH6371379281 Chem Impex International, Inc.

g-Glu-4-Abz-OH

  • MDL number MFCD00038802
  • Cas Number : 2643-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233-239
Synonym H-Glu(4-Abz-OH)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gamma-glu-4-abz-oh/3808?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gamma-glu-4-abz-oh/3808?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart