CH6371379288 Chem Impex International, Inc.

L-a-Aminobutyric acid tert-butyl ester hydrochloride

  • MDL number MFCD00237697
  • Cas Number : 53956-05-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 240-245
Synonym L-Abu-OtBu HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-a-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/3818?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-a-aminobutyric-acid-i-tert-i-butyl-ester-hydrochloride/3818?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart