CH6371379289 Chem Impex International, Inc.

a-Aminoisobutyric acid tert-butyl ester hydrochloride

  • MDL number MFCD08272284
  • Cas Number : 4512-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Aib-OtBu�
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/a-aminoisobutyric-acid-i-tert-i-butyl-ester-hydrochloride/3819?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/a-aminoisobutyric-acid-i-tert-i-butyl-ester-hydrochloride/3819?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart