CH6371379291 Chem Impex International, Inc.

Ala-D-Ala-OH

  • MDL number MFCD00066040
  • Cas Number : 3695-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Alanyl-D-alanine
Purity 0.97
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-d-ala-oh/3822?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-d-ala-oh/3822?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart