CH6371379292 Chem Impex International, Inc.

Ala-Lys-OH�

  • MDL number MFCD00058293
  • Cas Number : 68973-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Alanyl-L-lysine hydrochloride
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-lys-oh-font-size-2-face-symbol-middot-font-hcl/3823?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-lys-oh-font-size-2-face-symbol-middot-font-hcl/3823?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart