CH6371379294 Chem Impex International, Inc.

L-Alanine amide hydrobromide

  • MDL number MFCD00050707
  • Cas Number : 102029-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Ala-NH2 HBr
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-alanine-amide-hydrobromide/3825?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-alanine-amide-hydrobromide/3825?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart