CH6371379296 Chem Impex International, Inc.

L-Alanine 4-nitroanilide

  • MDL number MFCD00053640
  • Cas Number : 1668-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-alanine-4-nitroanilide/3828?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-alanine-4-nitroanilide/3828?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart