CH6371379298 Chem Impex International, Inc.

Ala-Ser-OH

  • MDL number MFCD00066028
  • Cas Number : 3303-41-1
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white to light redish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-ser-oh/3830?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-ser-oh/3830?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart