CH6371379300 Chem Impex International, Inc.

Ala-Val-OH

  • MDL number MFCD00066027
  • Cas Number : 3303-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144 - 150 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-val-oh/3835?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-val-oh/3835?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart