CH6371379302 Chem Impex International, Inc.

O-tert-Butyl-L-allo-threonine

  • MDL number MFCD00237373
  • Cas Number : 201353-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym allo-L-Thr(tBu)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-i-allo-i-threonine/3837?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-l-i-allo-i-threonine/3837?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart