CH6371379304 Chem Impex International, Inc.

L-allo-Threonine methyl ester hydrochloride

  • MDL number MFCD00237766
  • Cas Number : 79617-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym allo-L-Thr-OMe HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-l-allo-i-threonine-methyl-ester-hydrochloride/3839?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-l-allo-i-threonine-methyl-ester-hydrochloride/3839?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart