CH6371379305 Chem Impex International, Inc.

Nw-Methyl-L-arginine hydrochloride

  • MDL number MFCD01632059
  • Cas Number : 156706-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Arg(Me)-OH HCl
Purity 95
Storage temp. Store at 0-8
Physical Form White solid to thick paste
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-methyl-l-arginine-hydrochloride/3842?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-methyl-l-arginine-hydrochloride/3842?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart