CH6371379307 Chem Impex International, Inc.

Nw-(2,2,4,6,7-Pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine allyl ester hydrochloride

  • MDL number MFCD01632060
  • Cas Number : 257288-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Arg(Pbf)-OAll HCl
Purity 95
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-2-2-4-6-7-pentamethyldihydro-benzofuran-5-sulfonyl-l-arginine-allyl-ester-hydrochloride/3845?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-omega-sup-2-2-4-6-7-pentamethyldihydro-benzofuran-5-sulfonyl-l-arginine-allyl-ester-hydrochloride/3845?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart