CH6371379309 Chem Impex International, Inc.

H-Arg(Pmc)-OtBu (free base)

  • MDL number MFCD00236799
  • Cas Number : 169543-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119-123 °C
Purity 99
Storage temp. Store at = -4 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-arg-pmc-otbu-free-base/3847?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-arg-pmc-otbu-free-base/3847?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart