CH6371379312 Chem Impex International, Inc.

Arg-Tyr-OH AcOH

  • MDL number MFCD00190754
  • Molecular Weight : 397.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/arg-tyr-oh-font-size-2-face-symbol-middot-font-acoh/3851?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/arg-tyr-oh-font-size-2-face-symbol-middot-font-acoh/3851?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart