CH6371379314 Chem Impex International, Inc.

L-Asparagine 7-amido-4-methylcoumarin trifluoroacetate

  • MDL number MFCD00078916
  • Cas Number : 115047-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-118� C
Synonym L-Asn-AMC TFA
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-asparagine-7-amido-4-methylcoumarin-trifluoroacetate/3853?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-asparagine-7-amido-4-methylcoumarin-trifluoroacetate/3853?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart