CH6371379315 Chem Impex International, Inc.

L-Asparagine amide hydrochloride

  • MDL number MFCD00077102
  • Cas Number : 57471-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-asparagine-amide-hydrochloride/3854?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-asparagine-amide-hydrochloride/3854?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart