CH6371379316 Chem Impex International, Inc.

L-Asparagine tert-butyl ester

  • MDL number MFCD00190755
  • Cas Number : 25456-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Asn-OtBu
Purity 98
Storage temp. Store at 0-8
Physical Form White to off white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-asparagine-i-tert-i-butyl-ester/3855?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-asparagine-i-tert-i-butyl-ester/3855?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart