CH6371379320 Chem Impex International, Inc.

L-Aspartic acid beta-tert-butyl ester alpha-allyl ester hydrochloride

  • MDL number MFCD00798679
  • Cas Number : 218938-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-i-tert-i-butyl-ester-a-allyl-ester-hydrochloride/3860?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-i-tert-i-butyl-ester-a-allyl-ester-hydrochloride/3860?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart