CH6371379321 Chem Impex International, Inc.

L-Aspartic acid b-tert-butyl ester a-amide hydrochloride

  • MDL number MFCD00236805
  • Cas Number : 92786-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Asp(OtBu)-NH2 HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-i-tert-i-butyl-ester-a-amide-hydrochloride/3861?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-ß-i-tert-i-butyl-ester-a-amide-hydrochloride/3861?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart