CH6371379323 Chem Impex International, Inc.

L-Citrulline b-naphthylamide hydrobromide

  • MDL number MFCD00672746
  • Cas Number : 201988-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-137
Synonym L-Cit-betaNA HBr
Purity 99
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-citrulline-beta-naphthylamide-hydrobromide/3868?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-citrulline-beta-naphthylamide-hydrobromide/3868?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart