CH6371379324 Chem Impex International, Inc.

S-Benzyl-L-cysteine benzyl ester 4-toluenesulfonate salt

  • MDL number MFCD00038984
  • Cas Number : 73995-16-1
  • Molecular Weight : 473.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-164 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-benzyl-ester-4-toluenesulfonate-salt/3870?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-benzyl-ester-4-toluenesulfonate-salt/3870?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart