CH6371379325 Chem Impex International, Inc.

S-Benzyl-L-cysteine ethyl ester hydrochloride

  • MDL number MFCD00038909
  • Cas Number : 52844-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160 - 162 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-ethyl-ester-hydrochloride/3871?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-ethyl-ester-hydrochloride/3871?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart