CH6371379326 Chem Impex International, Inc.

S-Benzyl-L-cysteine 4-nitroanilide

  • MDL number MFCD00007302
  • Cas Number : 7436-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Cys(Bzl)-pNA
Purity 98
Storage temp. Store at 0-8
Physical Form Yellowish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-4-nitroanilide/3872?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-benzyl-l-cysteine-4-nitroanilide/3872?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart