CH6371379328 Chem Impex International, Inc.

S-Diphenylmethyl-L-cysteine

  • MDL number MFCD00065211
  • Cas Number : 5191-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-208 °C
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-diphenylmethyl-l-cysteine/3874?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-diphenylmethyl-l-cysteine/3874?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart