CH6371379330 Chem Impex International, Inc.

D-Ala-Ala-OH�

  • MDL number MFCD00066039

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-223
Synonym D-Alanyl-L-alanine hydrochloride
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-ala-ala-oh-font-size-2-face-symbol-middot-font-hcl/3878?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-ala-ala-oh-font-size-2-face-symbol-middot-font-hcl/3878?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart