CH6371379333 Chem Impex International, Inc.

D-Ala-Leu-OH

  • MDL number MFCD00066033
  • Cas Number : 67113-60-4
  • Molecular Weight : 202.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-122 °C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-ala-leu-oh/3882?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-ala-leu-oh/3882?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart