CH6371379334 Chem Impex International, Inc.

D-Ala-Phe-OH

  • MDL number MFCD00066030
  • Cas Number : 3061-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-ala-phe-oh/3883?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-ala-phe-oh/3883?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart