CH6371379338 Chem Impex International, Inc.

Nw-(2,2,4,6,7-Pentamethyldihydrobenzofuran)-5-sulfonyl-D-arginine

  • MDL number MFCD00236798
  • Cas Number : 200116-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Arg(Pbf)-OH
Purity 99.0
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-2-2-4-6-7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine/3888?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-2-2-4-6-7-pentamethyldihydrobenzofuran-5-sulfonyl-d-arginine/3888?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart