CH6371379339 Chem Impex International, Inc.

Nw-(4-Toluenesulfonyl)-D-arginine

  • MDL number MFCD00236800
  • Cas Number : 97233-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-112� C
Synonym D-Arg(Tos)-OH
Purity 97
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-4-toluenesulfonyl-d-arginine/3889?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-4-toluenesulfonyl-d-arginine/3889?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart