CH6371379340 Chem Impex International, Inc.

Ng-4-Methyltrityl-D-asparagine

  • MDL number MFCD00237159
  • Cas Number : 200203-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Asn(Mtt)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-4-methyltrityl-d-asparagine/3890?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-sup-4-methyltrityl-d-asparagine/3890?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart