CH6371379342 Chem Impex International, Inc.

D-Aspartic acid b-methyl ester hydrochloride

  • MDL number MFCD00236806
  • Cas Number : 21394-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Asp(OMe)-OH HCl
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-aspartic-acid-ß-methyl-ester-hydrochloride/3892?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-aspartic-acid-ß-methyl-ester-hydrochloride/3892?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart