CH6371379344 Chem Impex International, Inc.

D-Aspartic acid a-benzyl ester

  • MDL number MFCD00037289
  • Cas Number : 6367-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Asp-OBzl
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-aspartic-acid-a-benzyl-ester/3895?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-aspartic-acid-a-benzyl-ester/3895?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart