CH6371379347 Chem Impex International, Inc.

S-4-Methoxybenzyl-D-cysteine

  • MDL number MFCD00236946
  • Cas Number : 58290-34-9
  • Molecular Weight : 241.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218 - 222 °C
Purity 97
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-d-cysteine/3899?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-4-methoxybenzyl-d-cysteine/3899?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart