CH6371379355 Chem Impex International, Inc.

D-Glutamic acid a-tert-butyl ester hydrochloride

  • MDL number MFCD00237064
  • Cas Number : 200616-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Glu-OtBu HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-glutamic-acid-alpha-i-tert-i-butyl-ester-hydrochloride/3911?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-glutamic-acid-alpha-i-tert-i-butyl-ester-hydrochloride/3911?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart