CH6371379356 Chem Impex International, Inc.

Nim-Benzyl-D-histidine

  • MDL number MFCD00237096
  • Cas Number : 2022956-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-His(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-d-histidine/3912?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-benzyl-d-histidine/3912?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart