CH6371379358 Chem Impex International, Inc.

Nim-Trityl-D-histidine methyl ester hydrochloride

  • MDL number MFCD00237101
  • Cas Number : 200927-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-His(Trt)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-trityl-d-histidine-methyl-ester-hydrochloride/3914?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-im-sup-trityl-d-histidine-methyl-ester-hydrochloride/3914?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart