CH6371379359 Chem Impex International, Inc.

D-Homocitrulline

  • MDL number MFCD00237406
  • Cas Number : 121080-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-HomoCit-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-homocitrulline/3915?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-homocitrulline/3915?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart